The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024165.2 | NP_077084.1 | 12 - 13 | 1481 | 82 |
NR_027692.1 | 12 - 13 | 1470 | 82 | ||
XM_011514662.1 | XP_011512964.1 | 12 - 13 | 1398 | 82 | |
XM_011514663.1 | XP_011512965.1 | 12 - 13 | 1398 | 82 | |
XM_011514664.1 | XP_011512966.1 | 11 - 12 | 1302 | 82 | |
XM_017010939.1 | XP_016866428.1 | 10 - 11 | 1086 | 82 | |
GenBank mRNA | AF052205.1 | - | 12 - 13 | 1297 | 82 |
AK225913.1 | - | 11 - 12 | 1483 | 82 | |
AK295412.1 | - | 12 - 13 | 1340 | 82 | |
AK299162.1 | - | 9 - 10 | 969 | 82 | |
AK316499.1 | - | 12 - 13 | 1421 | 82 | |
BC008834.2 | - | 12 - 13 | 1449 | 82 |