The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174089.1 | NP_001167560.1 | 6 - 7 | 704 | 90 |
NM_001174090.1 | NP_001167561.1 | 6 - 7 | 854 | 90 | |
NM_032034.3 | NP_114423.1 | 5 - 6 | 696 | 90 | |
NR_135000.1 | 6 - 7 | 768 | 90 | ||
XM_005260856.4 | XP_005260913.1 | 6 - 7 | 1192 | 90 | |
XM_005260857.1 | XP_005260914.1 | 6 - 7 | 723 | 90 | |
XM_011529383.2 | XP_011527685.1 | 6 - 7 | 2299 | 90 | |
XM_011529384.1 | XP_011527686.1 | 6 - 7 | 941 | 90 | |
XM_011529385.1 | XP_011527687.1 | 6 - 7 | 768 | 90 | |
XM_017028093.1 | XP_016883582.1 | 6 - 7 | 1192 | 90 | |
XM_017028094.1 | XP_016883583.1 | 6 - 7 | 576 | 90 | |
XM_017028095.1 | XP_016883584.1 | 6 - 7 | 1047 | 90 | |
XM_017028096.1 | XP_016883585.1 | 7 - 8 | 953 | 90 | |
XM_017028097.1 | XP_016883586.1 | 6 - 7 | 1192 | 90 | |
GenBank mRNA | AF336127.1 | - | 5 - 6 | 696 | 90 |
AK091157.1 | - | 6 - 7 | 768 | 90 | |
AK296508.1 | - | 6 - 7 | 854 | 90 | |
AK296760.1 | - | 6 - 7 | 90 | ||
BC110540.1 | - | 5 - 6 | 90 | ||
BC110541.1 | - | 5 - 6 | 90 |