The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013432.4 | NP_038460.4 | 23 - 24 | 3753 | 63 |
XM_011517048.2 | XP_011515350.1 | 16 - 17 | 2852 | 63 | |
XM_011517049.2 | XP_011515351.1 | 16 - 17 | 2813 | 63 | |
GenBank mRNA | AB209126.1 | - | 14 - 15 | 5840 | 63 |
AK097406.1 | - | 2 - 3 | 1768 | 63 | |
BC008782.2 | - | 22 - 23 | 3725 | 63 |