The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017915.2 | NP_001017915.1 | 15 - 16 | 2304 | 71 |
NM_005541.4 | NP_005532.2 | 15 - 16 | 2301 | 71 | |
XM_017004004.1 | XP_016859493.1 | 15 - 16 | 1880 | 71 | |
GenBank mRNA | BC099920.1 | - | 15 - 16 | 1912 | 71 |
BC113580.1 | - | 15 - 16 | 1849 | 71 | |
BC113582.1 | - | 15 - 16 | 1849 | 71 | |
U50040.1 | - | 10 - 11 | 1169 | 71 | |
U50041.1 | - | 15 - 16 | 1913 | 71 | |
U53470.1 | - | 15 - 16 | 1922 | 71 | |
U84400.1 | - | 15 - 16 | 1873 | 71 | |
X98429.1 | - | 15 - 16 | 1929 | 71 |