The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017915.2 | NP_001017915.1 | 16 - 17 | 2411 | 77 |
NM_005541.4 | NP_005532.2 | 16 - 17 | 2408 | 77 | |
XM_017004004.1 | XP_016859493.1 | 16 - 17 | 1987 | 77 | |
GenBank mRNA | BC099920.1 | - | 16 - 17 | 2019 | 77 |
BC113580.1 | - | 16 - 17 | 1956 | 77 | |
BC113582.1 | - | 16 - 17 | 1956 | 77 | |
U50040.1 | - | 11 - 12 | 1276 | 77 | |
U50041.1 | - | 16 - 17 | 2020 | 77 | |
U53470.1 | - | 16 - 17 | 2029 | 77 | |
U57650.1 | - | 16 - 17 | 2408 | 77 | |
U84400.1 | - | 16 - 17 | 1980 | 77 | |
X98429.1 | - | 16 - 17 | 2036 | 77 |