The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017915.2 | NP_001017915.1 | 9 - 10 | 1540 | 93 |
NM_005541.4 | NP_005532.2 | 9 - 10 | 1537 | 93 | |
XM_017004004.1 | XP_016859493.1 | 9 - 10 | 1163 | 93 | |
GenBank mRNA | BC099920.1 | - | 9 - 10 | 1148 | 93 |
BC113580.1 | - | 9 - 10 | 1085 | 93 | |
BC113582.1 | - | 9 - 10 | 1085 | 93 | |
U50040.1 | - | 4 - 5 | 405 | 93 | |
U50041.1 | - | 9 - 10 | 1149 | 93 | |
U53470.1 | - | 9 - 10 | 1158 | 93 | |
U57650.1 | - | 9 - 10 | 1537 | 93 | |
U84400.1 | - | 9 - 10 | 1109 | 93 | |
X98429.1 | - | 9 - 10 | 1165 | 93 |