Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM15
Entrez Gene ID
8751
Gene Name
ADAM metallopeptidase domain 15
Gene Aliases
MDC15
Chromosome Location
Chr.1: 155051272 - 155062776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261464.1 NP_001248393.1 13 - 14 1563 71
NM_001261465.1 NP_001248394.1 13 - 14 1533 71
NM_001261466.1 NP_001248395.1 13 - 14 1485 71
NM_003815.4 NP_003806.3 13 - 14 1533 71
NM_207191.2 NP_997074.1 13 - 14 1533 71
NM_207194.2 NP_997077.1 13 - 14 1533 71
NM_207195.2 NP_997078.1 13 - 14 1533 71
NM_207196.2 NP_997079.1 13 - 14 1533 71
NM_207197.2 NP_997080.1 13 - 14 1533 71
NR_048577.1 13 - 14 1533 71
NR_048578.1 13 - 14 1533 71
NR_048579.1 13 - 14 1533 71
GenBank mRNA AB209157.1 - 12 - 13 3064 71
AK075498.1 - 5 - 6 607 71
AK295614.1 - 13 - 14 1505 71
AK296925.1 - 13 - 14 1484 71
AK297468.1 - 13 - 14 1406 71
AY518542.1 - 13 - 14 1418 71
AY560593.1 - 13 - 14 1519 71
AY560594.1 - 13 - 14 1519 71
AY560595.1 - 13 - 14 1519 71
AY560596.1 - 13 - 14 1519 71
AY560597.1 - 13 - 14 1519 71
AY560598.1 - 13 - 14 1519 71
AY560599.1 - 13 - 14 1519 71
AY560600.1 - 13 - 14 1519 71
AY560601.1 - 13 - 14 1519 71
AY576417.1 - 13 - 14 1519 71
BC014566.2 - 13 - 14 1499 71
BT009764.1 - 13 - 14 1418 71
U41767.1 - 13 - 14 1425 71
U46005.1 - 13 - 14 1418 71

View Full Product Details