Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM15
Entrez Gene ID
8751
Gene Name
ADAM metallopeptidase domain 15
Gene Aliases
MDC15
Chromosome Location
Chr.1: 155051272 - 155062776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261464.1 NP_001248393.1 17 - 18 2216 91
NM_001261465.1 NP_001248394.1 17 - 18 2186 91
NM_003815.4 NP_003806.3 17 - 18 2186 91
NM_207191.2 NP_997074.1 17 - 18 2186 91
NM_207194.2 NP_997077.1 17 - 18 2186 91
NM_207195.2 NP_997078.1 17 - 18 2186 91
NM_207196.2 NP_997079.1 17 - 18 2186 91
NM_207197.2 NP_997080.1 17 - 18 2186 91
NR_048577.1 17 - 18 2186 91
NR_048578.1 17 - 18 2186 91
NR_048579.1 17 - 18 2186 91
GenBank mRNA AB209157.1 - 16 - 17 3717 91
AK075498.1 - 9 - 10 1260 91
AK295614.1 - 17 - 18 2158 91
AK296925.1 - 17 - 18 2137 91
AY518542.1 - 17 - 18 2071 91
AY560593.1 - 17 - 18 2172 91
AY560594.1 - 17 - 18 2172 91
AY560595.1 - 17 - 18 2172 91
AY560596.1 - 17 - 18 2172 91
AY560597.1 - 17 - 18 2172 91
AY560598.1 - 17 - 18 2172 91
AY560599.1 - 17 - 18 2172 91
AY560600.1 - 17 - 18 2172 91
AY560601.1 - 17 - 18 2172 91
AY576417.1 - 17 - 18 2172 91
BC014566.2 - 17 - 18 2152 91
BT009764.1 - 17 - 18 2071 91
U41767.1 - 17 - 18 2078 91
U46005.1 - 17 - 18 2071 91

View Full Product Details