Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM15
Entrez Gene ID
8751
Gene Name
ADAM metallopeptidase domain 15
Gene Aliases
MDC15
Chromosome Location
Chr.1: 155051272 - 155062776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207196.2 NP_997079.1 20 - 21 2463 116
NM_207197.2 NP_997080.1 20 - 21 2466 116
GenBank mRNA AB209157.1 - 19 - 20 3994 116
AY560598.1 - 20 - 21 2449 116
AY560599.1 - 20 - 21 2452 116
BM971880.1 - 3 - 4 116

View Full Product Details