Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM15
Entrez Gene ID
8751
Gene Name
ADAM metallopeptidase domain 15
Gene Aliases
MDC15
Chromosome Location
Chr.1: 155051272 - 155062776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261465.1 NP_001248394.1 1 - 2 194 119
NM_003815.4 NP_003806.3 1 - 2 194 119
NM_207191.2 NP_997074.1 1 - 2 194 119
NM_207194.2 NP_997077.1 1 - 2 194 119
NM_207195.2 NP_997078.1 1 - 2 194 119
NM_207196.2 NP_997079.1 1 - 2 194 119
NM_207197.2 NP_997080.1 1 - 2 194 119
NR_048577.1 1 - 2 194 119
NR_048578.1 1 - 2 194 119
NR_048579.1 1 - 2 194 119
GenBank mRNA AK000667.1 - 1 - 2 180 119
AK075498.1 - 1 - 2 150 119
AK308172.1 - 1 - 2 117 119
AY518542.1 - 1 - 2 79 119
AY560593.1 - 1 - 2 180 119
AY560594.1 - 1 - 2 180 119
AY560595.1 - 1 - 2 180 119
AY560596.1 - 1 - 2 180 119
AY560597.1 - 1 - 2 180 119
AY560598.1 - 1 - 2 180 119
AY560599.1 - 1 - 2 180 119
AY560600.1 - 1 - 2 180 119
AY560601.1 - 1 - 2 180 119
AY576417.1 - 1 - 2 180 119
BC014566.2 - 1 - 2 160 119
BT009764.1 - 1 - 2 79 119
U41767.1 - 1 - 2 86 119
U46005.1 - 1 - 2 79 119

View Full Product Details