The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000722.3 | NP_000713.2 | 12 - 13 | 1492 | 111 |
XM_005250570.2 | XP_005250627.1 | 12 - 13 | 1599 | 111 | |
XM_005250572.2 | XP_005250629.1 | 12 - 13 | 1599 | 111 | |
XM_005250573.2 | XP_005250630.1 | 12 - 13 | 1596 | 111 | |
XM_005250574.2 | XP_005250631.1 | 12 - 13 | 1597 | 111 | |
XM_006716118.2 | XP_006716181.1 | 12 - 13 | 1601 | 111 | |
XM_006716119.3 | XP_006716182.1 | 12 - 13 | 3384 | 111 | |
XM_006716120.3 | XP_006716183.1 | 12 - 13 | 2826 | 111 | |
XM_011516570.2 | XP_011514872.1 | 12 - 13 | 1607 | 111 | |
XM_011516571.2 | XP_011514873.1 | 12 - 13 | 1602 | 111 | |
XM_011516572.2 | XP_011514874.1 | 12 - 13 | 1599 | 111 | |
XM_017012588.1 | XP_016868077.1 | 12 - 13 | 2825 | 111 | |
GenBank mRNA | BC117468.1 | - | 12 - 13 | 1176 | 111 |
BC117470.1 | - | 12 - 13 | 1176 | 111 | |
BG055272.1 | - | 1 - 2 | 122 | 111 |