The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001980.3 | NP_001971.2 | 3 - 4 | 379 | 144 |
NM_194356.2 | NP_919337.1 | 3 - 4 | 379 | 144 | |
XM_005253557.1 | XP_005253614.1 | 3 - 4 | 379 | 144 | |
XM_017018978.1 | XP_016874467.1 | 3 - 4 | 376 | 144 | |
XM_017018979.1 | XP_016874468.1 | 3 - 4 | 376 | 144 | |
XM_017018980.1 | XP_016874469.1 | 3 - 4 | 372 | 144 | |
XM_017018981.1 | XP_016874470.1 | 3 - 4 | 381 | 144 | |
XM_017018982.1 | XP_016874471.1 | 3 - 4 | 376 | 144 | |
XM_017018983.1 | XP_016874472.1 | 3 - 4 | 320 | 144 | |
XM_017018986.1 | XP_016874475.1 | 3 - 4 | 395 | 144 | |
XM_017018987.1 | XP_016874476.1 | 3 - 4 | 397 | 144 | |
XM_017018988.1 | XP_016874477.1 | 3 - 4 | 379 | 144 | |
GenBank mRNA | BC047496.1 | - | 3 - 4 | 371 | 144 |
D14582.1 | - | 3 - 4 | 299 | 144 | |
DB474363.1 | - | 3 - 4 | 379 | 144 |