The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025216.2 | NP_079492.2 | 2 - 3 | 835 | 66 |
XM_011511929.2 | XP_011510231.1 | 3 - 4 | 443 | 66 | |
GenBank mRNA | AB059569.1 | - | 2 - 3 | 377 | 66 |
AK024363.1 | - | 2 - 3 | 835 | 66 | |
AK315081.1 | - | 2 - 3 | 488 | 66 | |
AY009400.1 | - | 2 - 3 | 568 | 66 | |
BC034352.1 | - | 2 - 3 | 494 | 66 | |
BC040026.1 | - | 2 - 3 | 805 | 66 | |
BC052234.1 | - | 2 - 3 | 846 | 66 |