The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028292.1 | 5 - 6 | 1505 | 78 | |
NR_028293.1 | 4 - 5 | 1478 | 78 | ||
NR_028294.1 | 5 - 6 | 1574 | 78 | ||
GenBank mRNA | D86097.1 | - | 5 - 6 | 1505 | 78 |
D86098.1 | - | 4 - 5 | 1478 | 78 | |
X83863.1 | - | 5 - 6 | 1550 | 78 |