Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
Chromosome Location
Chr.1: 59296405 - 59762730 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244714.1 NP_001231643.1 9 - 10 1068 68
NM_001278224.1 NP_001265153.1 5 - 6 587 68
NM_018291.3 NP_060761.3 11 - 12 1431 68
NR_103473.1 12 - 13 1389 68
XM_011541735.1 XP_011540037.1 11 - 12 1250 68
XM_017001645.1 XP_016857134.1 11 - 12 1355 68
XM_017001646.1 XP_016857135.1 11 - 12 1332 68
XM_017001647.1 XP_016857136.1 11 - 12 1460 68
XM_017001650.1 XP_016857139.1 13 - 14 1623 68
XM_017001651.1 XP_016857140.1 13 - 14 1479 68
XM_017001652.1 XP_016857141.1 11 - 12 1400 68
XM_017001653.1 XP_016857142.1 11 - 12 1332 68
XM_017001666.1 XP_016857155.1 9 - 10 1146 68
XM_017001667.1 XP_016857156.1 9 - 10 1200 68
XM_017001672.1 XP_016857161.1 10 - 11 1288 68
XM_017001673.1 XP_016857162.1 10 - 11 1345 68
XM_017001674.1 XP_016857163.1 10 - 11 1204 68
XM_017001676.1 XP_016857165.1 10 - 11 1348 68
GenBank mRNA AK001848.1 - 5 - 6 461 68
AK054842.1 - 9 - 10 1059 68
AK315649.1 - 11 - 12 1316 68
BC000610.2 - 5 - 6 562 68
BC014947.1 - 11 - 12 1316 68

View Full Product Details