The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000754.3 | NP_000745.1 | 4 - 5 | 733 | 63 |
NM_001135161.1 | NP_001128633.1 | 4 - 5 | 691 | 63 | |
NM_001135162.1 | NP_001128634.1 | 4 - 5 | 708 | 63 | |
NM_007310.2 | NP_009294.1 | 2 - 3 | 464 | 63 | |
XM_011529886.1 | XP_011528188.1 | 4 - 5 | 638 | 63 | |
XM_017028594.1 | XP_016884083.1 | 4 - 5 | 957 | 63 | |
XM_017028595.1 | XP_016884084.1 | 4 - 5 | 901 | 63 | |
GenBank mRNA | AK130031.1 | - | 5 - 6 | 513 | 63 |
AK290440.1 | - | 4 - 5 | 671 | 63 | |
BC000419.2 | - | 4 - 5 | 63 | ||
BC005867.2 | - | 4 - 5 | 63 | ||
BC011935.2 | - | 3 - 4 | 576 | 63 | |
BC100018.1 | - | 4 - 5 | 723 | 63 | |
CR456422.1 | - | 3 - 4 | 63 | ||
CR456997.1 | - | 2 - 3 | 484 | 63 | |
EF103918.1 | - | 2 - 3 | 225 | 63 | |
EF103922.1 | - | 3 - 4 | 513 | 63 | |
FJ224345.1 | - | 4 - 5 | 63 | ||
FJ410131.1 | - | 2 - 3 | 334 | 63 | |
FJ410132.1 | - | 2 - 3 | 484 | 63 | |
M58525.1 | - | 4 - 5 | 63 | ||
M65212.1 | - | 3 - 4 | 562 | 63 |