The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002454.2 | NP_002445.2 | 7 - 8 | 1198 | 87 |
NM_024010.2 | NP_076915.2 | 7 - 8 | 1172 | 87 | |
NR_134480.1 | 7 - 8 | 1184 | 87 | ||
NR_134482.1 | 6 - 7 | 1044 | 87 | ||
GenBank mRNA | AF025794.1 | - | 7 - 8 | 1140 | 87 |
AF121214.1 | - | 7 - 8 | 1172 | 87 | |
AK308442.1 | - | 8 - 9 | 1128 | 87 | |
BC054816.1 | - | 7 - 8 | 1113 | 87 | |
BC109216.1 | - | 7 - 8 | 1172 | 87 |