The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014676.2 | NP_055491.1 | 17 - 18 | 2962 | 72 |
GenBank mRNA | AF315592.1 | - | 17 - 18 | 2944 | 72 |
AK291779.1 | - | 17 - 18 | 2931 | 72 | |
AK294477.1 | - | 15 - 16 | 2672 | 72 | |
BC013398.2 | - | 18 - 19 | 2919 | 72 |