Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN1
Entrez Gene ID
9378
Gene Name
neurexin 1
Gene Aliases
Hs.22998, PTHSL2, SCZD17
Chromosome Location
Chr.2: 49918503 - 51032536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135659.1 NP_001129131.1 13 - 14 3969 107
NM_004801.4 NP_004792.1 12 - 13 3849 107
XM_005264642.3 XP_005264699.1 12 - 13 3870 107
XM_005264643.3 XP_005264700.1 11 - 12 3825 107
XM_006712137.3 XP_006712200.1 12 - 13 3870 107
XM_006712140.3 XP_006712203.2 12 - 13 3870 107
XM_011533167.2 XP_011531469.1 12 - 13 3870 107
XM_011533171.2 XP_011531473.1 12 - 13 3849 107
XM_011533174.2 XP_011531476.1 11 - 12 3825 107
XM_011533175.2 XP_011531477.1 10 - 11 3813 107
XM_011533177.2 XP_011531479.1 12 - 13 3870 107
XM_011533180.2 XP_011531482.1 12 - 13 3870 107
XM_017005303.1 XP_016860792.1 12 - 13 3870 107
XM_017005304.1 XP_016860793.1 12 - 13 3867 107
XM_017005305.1 XP_016860794.1 12 - 13 3870 107
XM_017005306.1 XP_016860795.1 11 - 12 3858 107
XM_017005307.1 XP_016860796.1 11 - 12 3852 107
XM_017005308.1 XP_016860797.1 12 - 13 3849 107
XM_017005310.1 XP_016860799.1 10 - 11 3840 107
XM_017005311.1 XP_016860800.1 11 - 12 3825 107
XM_017005312.1 XP_016860801.1 12 - 13 3846 107
XM_017005313.1 XP_016860802.1 12 - 13 3849 107
XM_017005314.1 XP_016860803.1 9 - 10 3810 107
XM_017005316.1 XP_016860805.1 10 - 11 3807 107
XM_017005317.1 XP_016860806.1 11 - 12 3837 107
XM_017005321.1 XP_016860810.1 12 - 13 3870 107
XM_017005322.1 XP_016860811.1 12 - 13 3870 107
XM_017005326.1 XP_016860815.1 11 - 12 3831 107
XM_017005327.1 XP_016860816.1 11 - 12 3825 107
XM_017005329.1 XP_016860818.1 12 - 13 3870 107
XM_017005334.1 XP_016860823.1 8 - 9 1691 107
GenBank mRNA AB011150.2 - 10 - 11 2531 107
AB035356.1 - 11 - 12 2564 107
BC150247.1 - 10 - 11 2552 107
EF539882.1 - 13 - 14 3832 107

View Full Product Details