Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN1
Entrez Gene ID
9378
Gene Name
neurexin 1
Gene Aliases
Hs.22998, PTHSL2, SCZD17
Chromosome Location
Chr.2: 49918503 - 51032536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135659.1 NP_001129131.1 17 - 18 4841 57
NM_004801.4 NP_004792.1 16 - 17 4721 57
XM_005264642.3 XP_005264699.1 16 - 17 4742 57
XM_005264643.3 XP_005264700.1 15 - 16 4697 57
XM_006712137.3 XP_006712200.1 16 - 17 4742 57
XM_011533167.2 XP_011531469.1 16 - 17 4742 57
XM_011533171.2 XP_011531473.1 16 - 17 4721 57
XM_011533172.2 XP_011531474.1 15 - 16 4715 57
XM_011533174.2 XP_011531476.1 15 - 16 4697 57
XM_011533175.2 XP_011531477.1 14 - 15 4685 57
XM_011533177.2 XP_011531479.1 16 - 17 4742 57
XM_011533178.2 XP_011531480.1 13 - 14 4652 57
XM_011533180.2 XP_011531482.1 16 - 17 4742 57
XM_011533183.1 XP_011531485.1 10 - 11 2424 57
XM_017005312.1 XP_016860801.1 16 - 17 4718 57
XM_017005313.1 XP_016860802.1 16 - 17 4721 57
XM_017005315.1 XP_016860804.1 15 - 16 4715 57
XM_017005317.1 XP_016860806.1 15 - 16 4709 57
XM_017005319.1 XP_016860808.1 15 - 16 4694 57
XM_017005320.1 XP_016860809.1 15 - 16 4694 57
XM_017005323.1 XP_016860812.1 12 - 13 4655 57
XM_017005324.1 XP_016860813.1 15 - 16 4715 57
XM_017005325.1 XP_016860814.1 15 - 16 4715 57
XM_017005326.1 XP_016860815.1 15 - 16 4703 57
XM_017005327.1 XP_016860816.1 15 - 16 4697 57
XM_017005328.1 XP_016860817.1 14 - 15 4670 57
XM_017005330.1 XP_016860819.1 12 - 13 4655 57
XM_017005331.1 XP_016860820.1 12 - 13 4640 57
XM_017005332.1 XP_016860821.1 11 - 12 4610 57
XM_017005333.1 XP_016860822.1 11 - 12 4610 57
GenBank mRNA AB011150.2 - 14 - 15 3403 57
AB035356.1 - 15 - 16 3436 57
AK093260.1 - 10 - 11 2424 57
BC150247.1 - 14 - 15 3424 57
EF539882.1 - 17 - 18 4704 57

View Full Product Details