Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TTTY10
- Entrez Gene ID
-
246119
- Gene Name
- testis-specific transcript, Y-linked 10 (non-protein coding)
- Gene Aliases
-
NCRNA00133, TTY10
- Chromosome Location
-
Chr.Y: 20465668 - 20519228 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001542.1 |
|
5 - 6 |
1476 |
71 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF332239.3 |
- |
5 - 6 |
1476 |
|
Chr. Y - 20465668 - 20519228 [-] on Build GRCh38 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
lysine demethylase 5D |
KDM5D |
HY, HYA, JARID1D, SMCY |
-720.7kb
|
Sense |
Chr. - 19692495 - 19745347 on Build GRCh38 |
Downstream |
eukaryotic translation initiation factor 1A, Y-linked |
EIF1AY |
eIF-4C |
109.0kb
|
Anti-sense |
Chr. - 20575711 - 20593154 on Build GRCh38 |
View Full Product Details