The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278622.1 | NP_001265551.1 | 10 - 11 | 1426 | 123 |
NM_002873.1 | NP_002864.1 | 9 - 10 | 1531 | 123 | |
NM_133338.2 | NP_579916.1 | 11 - 12 | 1427 | 123 | |
NM_133339.2 | NP_579917.1 | 8 - 9 | 984 | 123 | |
NM_133340.2 | NP_579918.1 | 8 - 9 | 1017 | 123 | |
NM_133341.2 | NP_579919.1 | 8 - 9 | 943 | 123 | |
NM_133342.2 | NP_579920.1 | 10 - 11 | 1404 | 123 | |
NM_133343.1 | NP_579921.1 | 10 - 11 | 1369 | 123 | |
NM_133344.2 | NP_579922.1 | 9 - 10 | 1186 | 123 | |
XM_017009679.1 | XP_016865168.1 | 10 - 11 | 1246 | 123 | |
XM_017009680.1 | XP_016865169.1 | 9 - 10 | 1300 | 123 | |
XM_017009681.1 | XP_016865170.1 | 9 - 10 | 1158 | 123 | |
GenBank mRNA | AF017748.1 | - | 9 - 10 | 1156 | 123 |
AF076838.1 | - | 9 - 10 | 1531 | 123 | |
AF098533.1 | - | 8 - 9 | 963 | 123 | |
AF098534.1 | - | 8 - 9 | 889 | 123 | |
AF112263.1 | - | 8 - 9 | 1008 | 123 | |
AF126424.1 | - | 11 - 12 | 1426 | 123 | |
AJ001642.1 | - | 9 - 10 | 1195 | 123 | |
AJ004977.1 | - | 9 - 10 | 1191 | 123 | |
AK292487.1 | - | 10 - 11 | 1404 | 123 | |
AL122068.1 | - | 10 - 11 | 1369 | 123 | |
BC032304.1 | - | 8 - 9 | 997 | 123 | |
BX537441.1 | - | 10 - 11 | 1426 | 123 |