The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278622.1 | NP_001265551.1 | 8 - 9 | 1234 | 108 |
NM_002873.1 | NP_002864.1 | 7 - 8 | 1339 | 108 | |
NM_133338.2 | NP_579916.1 | 9 - 10 | 1235 | 108 | |
NM_133339.2 | NP_579917.1 | 6 - 7 | 792 | 108 | |
NM_133340.2 | NP_579918.1 | 6 - 7 | 825 | 108 | |
NM_133341.2 | NP_579919.1 | 6 - 7 | 751 | 108 | |
NM_133342.2 | NP_579920.1 | 8 - 9 | 1212 | 108 | |
NM_133343.1 | NP_579921.1 | 8 - 9 | 1177 | 108 | |
NM_133344.2 | NP_579922.1 | 7 - 8 | 994 | 108 | |
XM_017009679.1 | XP_016865168.1 | 8 - 9 | 1054 | 108 | |
XM_017009680.1 | XP_016865169.1 | 7 - 8 | 1108 | 108 | |
XM_017009681.1 | XP_016865170.1 | 7 - 8 | 966 | 108 | |
GenBank mRNA | AF017748.1 | - | 7 - 8 | 964 | 108 |
AF076838.1 | - | 7 - 8 | 1339 | 108 | |
AF085736.2 | - | 6 - 7 | 792 | 108 | |
AF098533.1 | - | 6 - 7 | 771 | 108 | |
AF098534.1 | - | 6 - 7 | 697 | 108 | |
AF112263.1 | - | 6 - 7 | 816 | 108 | |
AF126424.1 | - | 9 - 10 | 1234 | 108 | |
AJ001642.1 | - | 7 - 8 | 1003 | 108 | |
AJ004977.1 | - | 7 - 8 | 999 | 108 | |
AK292487.1 | - | 8 - 9 | 1212 | 108 | |
AL122068.1 | - | 8 - 9 | 1177 | 108 | |
BC032304.1 | - | 6 - 7 | 805 | 108 | |
BX537441.1 | - | 8 - 9 | 1234 | 108 |