The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031885.3 | NP_114091.3 | 15 - 16 | 2144 | 134 |
XM_005256080.2 | XP_005256137.1 | 15 - 16 | 2143 | 134 | |
GenBank mRNA | AB208905.1 | - | 10 - 11 | 5441 | 134 |
AF342736.1 | - | 15 - 16 | 2331 | 134 | |
AK027635.1 | - | 15 - 16 | 2072 | 134 | |
AK127290.1 | - | 2 - 3 | 3398 | 134 | |
AK289604.1 | - | 15 - 16 | 2034 | 134 | |
AL834176.1 | - | 3 - 4 | 1492 | 134 | |
BC014140.2 | - | 15 - 16 | 2034 | 134 |