The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031885.3 | NP_114091.3 | 1 - 2 | 352 | 84 |
XM_005256080.2 | XP_005256137.1 | 1 - 2 | 351 | 84 | |
GenBank mRNA | AF342736.1 | - | 1 - 2 | 539 | 84 |
AK027635.1 | - | 1 - 2 | 280 | 84 | |
AK289604.1 | - | 1 - 2 | 242 | 84 | |
AK309068.1 | - | 1 - 2 | 252 | 84 | |
BC014140.2 | - | 1 - 2 | 242 | 84 | |
DA437930.1 | - | 1 - 2 | 355 | 84 |