The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031885.3 | NP_114091.3 | 5 - 6 | 842 | 113 |
XM_005256080.2 | XP_005256137.1 | 5 - 6 | 841 | 113 | |
GenBank mRNA | AB208905.1 | - | 5 - 6 | 622 | 113 |
AF342736.1 | - | 5 - 6 | 1029 | 113 | |
AK027635.1 | - | 5 - 6 | 770 | 113 | |
AK289604.1 | - | 5 - 6 | 732 | 113 | |
AK309068.1 | - | 5 - 6 | 742 | 113 | |
BC014140.2 | - | 5 - 6 | 732 | 113 |