The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018072.5 | NP_060542.4 | 29 - 30 | 4229 | 159 |
GenBank mRNA | AK098212.1 | - | 4 - 5 | 500 | 159 |
AK125663.1 | - | 11 - 12 | 1446 | 159 | |
AK304129.1 | - | 9 - 10 | 1306 | 159 | |
AK316413.1 | - | 9 - 10 | 1200 | 159 | |
BC065205.1 | - | 7 - 8 | 965 | 159 | |
BC130000.1 | - | 14 - 15 | 2153 | 159 | |
BC150614.1 | - | 28 - 29 | 4099 | 159 |