The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018072.5 | NP_060542.4 | 32 - 33 | 4756 | 89 |
GenBank mRNA | AK125663.1 | - | 14 - 15 | 1973 | 89 |
AK304129.1 | - | 12 - 13 | 1833 | 89 | |
AK316413.1 | - | 12 - 13 | 1727 | 89 | |
BC035302.1 | - | 3 - 4 | 331 | 89 | |
BC065205.1 | - | 10 - 11 | 1492 | 89 | |
BC130000.1 | - | 17 - 18 | 2680 | 89 | |
BC150614.1 | - | 31 - 32 | 4626 | 89 |