Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHOX
Entrez Gene ID
6473
Gene Name
short stature homeobox
Gene Aliases
GCFX, PHOG, SHOXY, SS
Chromosome Location
Chr.Y: 624344 - 659411 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000451.3 NP_000442.1 2 - 3 961 85
NM_006883.2 NP_006874.1 2 - 3 961 85
GenBank mRNA U89331.1 - 2 - 3 963 85
Y11535.1 - 1 - 2 361 85
Y11536.1 - 1 - 2 361 85

View Full Product Details