The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018968.3 | NP_061841.2 | 15 - 16 | 1511 | 90 |
XM_011510366.2 | XP_011508668.1 | 13 - 14 | 1548 | 90 | |
XM_017004361.1 | XP_016859850.1 | 15 - 16 | 1473 | 90 | |
XM_017004363.1 | XP_016859852.1 | 15 - 16 | 1660 | 90 | |
XM_017004364.1 | XP_016859853.1 | 12 - 13 | 4801 | 90 | |
XM_017004367.1 | XP_016859856.1 | 16 - 17 | 1549 | 90 | |
XM_017004373.1 | XP_016859862.1 | 10 - 11 | 2115 | 90 | |
GenBank mRNA | AJ003029.1 | - | 15 - 16 | 1513 | 90 |
BC125251.1 | - | 9 - 10 | 1059 | 90 | |
BG208326.1 | - | 1 - 2 | 97 | 90 |