The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166107.1 | NP_001159579.1 | 7 - 8 | 1385 | 113 |
NM_005518.3 | NP_005509.1 | 8 - 9 | 1511 | 113 | |
XM_011541313.1 | XP_011539615.1 | 7 - 8 | 1539 | 113 | |
GenBank mRNA | AK301594.1 | - | 7 - 8 | 1168 | 113 |
AK302276.1 | - | 7 - 8 | 1354 | 113 | |
AK303777.1 | - | 7 - 8 | 1358 | 113 | |
AK313590.1 | - | 8 - 9 | 1470 | 113 | |
BC044217.1 | - | 8 - 9 | 1464 | 113 | |
CR456850.1 | - | 9 - 10 | 1423 | 113 | |
U12789.1 | - | 7 - 8 | 990 | 113 | |
X83618.1 | - | 8 - 9 | 1474 | 113 |