The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018946.3 | NP_061819.2 | 4 - 5 | 734 | 65 |
XM_011518787.2 | XP_011517089.1 | 4 - 5 | 490 | 65 | |
XM_011518788.2 | XP_011517090.1 | 2 - 3 | 241 | 65 | |
XM_017014811.1 | XP_016870300.1 | 3 - 4 | 435 | 65 | |
GenBank mRNA | AF257466.1 | - | 4 - 5 | 602 | 65 |
AK001659.1 | - | 4 - 5 | 672 | 65 | |
AK091184.1 | - | 2 - 3 | 2528 | 65 | |
AK316608.1 | - | 4 - 5 | 694 | 65 | |
AL526281.3 | - | 4 - 5 | 734 | 65 | |
BC000008.2 | - | 4 - 5 | 680 | 65 | |
BC019315.1 | - | 4 - 5 | 662 | 65 | |
BQ446501.1 | - | 2 - 3 | 65 | ||
FJ224349.1 | - | 4 - 5 | 602 | 65 |