Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
Chromosome Location
Chr.1: 27872531 - 27886685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105556.2 NP_001099026.1 2 - 3 267 86
NM_001286113.1 NP_001273042.1 2 - 3 267 86
NM_001286115.1 NP_001273044.1 2 - 3 267 86
NM_004848.3 NP_004839.2 2 - 3 267 86
XM_006711050.1 XP_006711113.1 2 - 3 261 86
GenBank mRNA AK303090.1 - 2 - 3 229 86
AK303141.1 - 2 - 3 263 86
BC031655.1 - 2 - 3 235 86
BC132692.1 - 2 - 3 266 86
BC133049.1 - 2 - 3 266 86
BM146655.1 - 2 - 3 267 86

View Full Product Details