Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHPF2
Entrez Gene ID
54480
Gene Name
chondroitin polymerizing factor 2
Gene Aliases
CSGLCA-T, CSGlcAT, ChSy-3, chPF-2
Chromosome Location
Chr.7: 151232489 - 151238827 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284295.1 NP_001271224.1 2 - 3 517 88
NM_019015.2 NP_061888.1 1 - 2 1788 88
GenBank mRNA AB037823.1 - 1 - 2 1778 88
AB095812.1 - 1 - 2 1778 88
AK056331.1 - 2 - 3 517 88
AY358407.1 - 1 - 2 597 88
BC064509.1 - 1 - 2 747 88

View Full Product Details