Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL3
Entrez Gene ID
94120
Gene Name
synaptotagmin like 3
Gene Aliases
SLP3
Chromosome Location
Chr.6: 158644881 - 158766298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009991.3 NP_001009991.2 15 - 16 1697 75
NM_001242384.1 NP_001229313.1 17 - 18 1917 75
NM_001242394.1 NP_001229323.1 16 - 17 2309 75
NM_001242395.1 NP_001229324.1 14 - 15 2105 75
NM_001318745.1 NP_001305674.1 13 - 14 1631 75
XM_005267215.4 XP_005267272.1 16 - 17 2407 75
XM_005267218.4 XP_005267275.1 16 - 17 1718 75
XM_005267222.4 XP_005267279.1 8 - 9 989 75
XM_006715605.3 XP_006715668.1 17 - 18 2499 75
XM_006715606.3 XP_006715669.1 17 - 18 2420 75
XM_006715611.3 XP_006715674.1 16 - 17 2022 75
XM_011536254.2 XP_011534556.1 17 - 18 2188 75
XM_011536255.2 XP_011534557.1 7 - 8 1244 75
XM_017011496.1 XP_016866985.1 16 - 17 2588 75
XM_017011497.1 XP_016866986.1 17 - 18 1925 75
XM_017011498.1 XP_016866987.1 13 - 14 1738 75
GenBank mRNA AK055750.1 - 13 - 14 1630 75
AK298712.1 - 17 - 18 1877 75
AK304191.1 - 8 - 9 1174 75
BC085612.1 - 14 - 15 1643 75
BC100830.1 - 15 - 16 1698 75
BC100831.1 - 15 - 16 1698 75

View Full Product Details