The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270952.1 | NP_001257881.1 | 3 - 4 | 333 | 135 |
NM_006002.4 | NP_005993.1 | 3 - 4 | 255 | 135 | |
XM_011535212.1 | XP_011533514.1 | 3 - 4 | 255 | 135 | |
XM_011535213.2 | XP_011533515.1 | 2 - 3 | 236 | 135 | |
XM_011535214.2 | XP_011533516.1 | 3 - 4 | 323 | 135 | |
XM_017020725.1 | XP_016876214.1 | 3 - 4 | 255 | 135 | |
XM_017020726.1 | XP_016876215.1 | 3 - 4 | 255 | 135 | |
XM_017020727.1 | XP_016876216.1 | 2 - 3 | 230 | 135 | |
GenBank mRNA | AK313665.1 | - | 3 - 4 | 245 | 135 |
BC018125.2 | - | 3 - 4 | 214 | 135 | |
BQ427804.1 | - | 3 - 4 | 269 | 135 | |
BT019359.1 | - | 3 - 4 | 184 | 135 | |
CR456855.1 | - | 3 - 4 | 184 | 135 | |
DA820150.1 | - | 3 - 4 | 255 | 135 | |
M30496.1 | - | 3 - 4 | 204 | 135 |