The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291970.1 | NP_001278899.1 | 10 - 11 | 1554 | 97 |
GenBank mRNA | BC015742.1 | - | 10 - 11 | 1398 | 97 |