The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000600.4 | NP_000591.1 | 1 - 2 | 139 | 60 |
XM_011515390.2 | XP_011513692.1 | 2 - 3 | 472 | 60 | |
GenBank mRNA | A09363.1 | - | 1 - 2 | 104 | 60 |
AK301141.1 | - | 1 - 2 | 134 | 60 | |
BC015511.1 | - | 1 - 2 | 67 | 60 | |
HY076725.1 | - | 1 - 2 | 140 | 60 | |
M14584.1 | - | 1 - 2 | 81 | 60 | |
M18403.1 | - | 1 - 2 | 80 | 60 | |
M29150.1 | - | 1 - 2 | 53 | 60 | |
M54894.1 | - | 1 - 2 | 68 | 60 | |
X04430.1 | - | 1 - 2 | 104 | 60 | |
X04602.1 | - | 1 - 2 | 51 | 60 |