The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160305.2 | NP_001153777.1 | 7 - 8 | 1172 | 66 |
NM_024860.3 | NP_079136.2 | 8 - 9 | 1100 | 66 | |
NR_134583.1 | 8 - 9 | 1131 | 66 | ||
GenBank mRNA | AK024801.1 | - | 8 - 9 | 1056 | 66 |
AK127136.1 | - | 4 - 5 | 2577 | 66 | |
AK290495.1 | - | 7 - 8 | 1134 | 66 | |
BC022451.1 | - | 7 - 8 | 1134 | 66 | |
BC110433.1 | - | 8 - 9 | 1115 | 66 |