Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
Chromosome Location
Chr.11: 6497296 - 6572025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144666.2 NP_653267.2 37 - 38 13096 92
GenBank mRNA AB290171.1 - 24 - 25 9796 92
AK057314.1 - 4 - 5 1615 92
AK128064.1 - 5 - 6 2738 92
AL137690.1 - 2 - 3 693 92
AL834222.1 - 4 - 5 2556 92
BC082757.1 - 6 - 7 1972 92
BC117301.1 - 4 - 5 1440 92
BC117303.1 - 4 - 5 1440 92
BC143910.1 - 4 - 5 1410 92

View Full Product Details