The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166280.1 | NP_001159752.1 | 13 - 14 | 1802 | 82 |
NM_001166281.1 | NP_001159753.1 | 12 - 13 | 1772 | 82 | |
NM_005592.3 | NP_005583.1 | 14 - 15 | 2060 | 82 | |
XM_005251994.3 | XP_005252051.1 | 15 - 16 | 2672 | 82 | |
XM_005251995.3 | XP_005252052.1 | 14 - 15 | 2647 | 82 | |
XM_005251996.3 | XP_005252053.1 | 13 - 14 | 2616 | 82 | |
XM_011518708.2 | XP_011517010.1 | 6 - 7 | 805 | 82 | |
XM_017014734.1 | XP_016870223.1 | 14 - 15 | 2408 | 82 | |
GenBank mRNA | AF006464.1 | - | 14 - 15 | 1972 | 82 |
BC109098.1 | - | 13 - 14 | 1714 | 82 | |
BC109099.1 | - | 12 - 13 | 1684 | 82 |