The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318732.1 | NP_001305661.1 | 9 - 10 | 1912 | 125 |
NM_024089.2 | NP_076994.2 | 8 - 9 | 1769 | 125 | |
GenBank mRNA | AK074974.1 | - | 9 - 10 | 1830 | 125 |
AK222567.1 | - | 8 - 9 | 1691 | 125 | |
AK225103.1 | - | 8 - 9 | 1692 | 125 | |
AY358959.1 | - | 8 - 9 | 1670 | 125 | |
BC001297.1 | - | 8 - 9 | 1722 | 125 |