The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015285.2 | NP_056100.2 | 20 - 21 | 3518 | 96 |
NM_052834.2 | NP_443066.2 | 19 - 20 | 3419 | 96 | |
XM_006722431.2 | XP_006722494.1 | 20 - 21 | 3450 | 96 | |
XM_011525888.1 | XP_011524190.1 | 20 - 21 | 3560 | 96 | |
XM_017025682.1 | XP_016881171.1 | 19 - 20 | 3324 | 96 | |
XM_017025683.1 | XP_016881172.1 | 10 - 11 | 1911 | 96 | |
GenBank mRNA | AB011113.2 | - | 20 - 21 | 3481 | 96 |
AF188125.1 | - | 18 - 19 | 3208 | 96 | |
AY099325.1 | - | 19 - 20 | 3307 | 96 | |
BC050352.1 | - | 6 - 7 | 679 | 96 | |
BC131558.1 | - | 8 - 9 | 1282 | 96 | |
BC150282.1 | - | 20 - 21 | 3481 | 96 |