The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163280.1 | NP_001156752.1 | 5 - 6 | 989 | 122 |
NM_014500.4 | NP_055315.2 | 4 - 5 | 740 | 122 | |
XM_005262404.3 | XP_005262461.1 | 5 - 6 | 1084 | 122 | |
GenBank mRNA | AB209303.1 | - | 5 - 6 | 989 | 122 |
AK225870.1 | - | 4 - 5 | 687 | 122 | |
AK299403.1 | - | 4 - 5 | 692 | 122 | |
BC009896.2 | - | 4 - 5 | 740 | 122 | |
BT006886.1 | - | 4 - 5 | 567 | 122 | |
U76992.1 | - | 4 - 5 | 624 | 122 |