Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CROCC
Entrez Gene ID
9696
Gene Name
ciliary rootlet coiled-coil, rootletin
Gene Aliases
ROLT
Chromosome Location
Chr.1: 16914082 - 16972979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014675.4 NP_055490.4 8 - 9 1023 83
XM_006711058.3 XP_006711121.1 7 - 8 814 83
XM_011542468.1 XP_011540770.1 8 - 9 1000 83
XM_011542470.2 XP_011540772.1 8 - 9 1000 83
XM_017002913.1 XP_016858402.1 8 - 9 1000 83
XM_017002914.1 XP_016858403.1 8 - 9 1000 83
XM_017002915.1 XP_016858404.1 8 - 9 1000 83
GenBank mRNA AB007914.2 - 6 - 7 662 83
BC126911.1 - 5 - 6 544 83
BC126912.1 - 5 - 6 544 83

View Full Product Details