The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145725.1 | NP_001139197.1 | 2 - 3 | 89 | 134 |
NM_017816.2 | NP_060286.1 | 2 - 3 | 187 | 134 | |
XM_011513505.1 | XP_011511807.1 | 2 - 3 | 489 | 134 | |
GenBank mRNA | AI418609.1 | - | 2 - 3 | 187 | 134 |
AK000432.1 | - | 2 - 3 | 134 | 134 | |
AK290750.1 | - | 2 - 3 | 164 | 134 | |
AL136750.1 | - | 2 - 3 | 89 | 134 | |
BC015796.2 | - | 2 - 3 | 149 | 134 |