The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014777.3 | NP_055592.2 | 1 - 2 | 141 | 121 |
XM_005273360.3 | XP_005273417.1 | 1 - 2 | 110 | 121 | |
GenBank mRNA | AK291013.1 | - | 1 - 2 | 141 | 121 |
BC059381.1 | - | 1 - 2 | 116 | 121 | |
BC114963.1 | - | 1 - 2 | 63 | 121 | |
D50923.2 | - | 1 - 2 | 123 | 121 | |
DC369323.1 | - | 1 - 2 | 141 | 121 |