The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025248.2 | NP_079524.2 | 10 - 11 | 2251 | 98 |
XM_017025169.1 | XP_016880658.1 | 11 - 12 | 2563 | 98 | |
XM_017025170.1 | XP_016880659.1 | 11 - 12 | 2418 | 98 | |
XM_017025171.1 | XP_016880660.1 | 11 - 12 | 2418 | 98 | |
XM_017025172.1 | XP_016880661.1 | 10 - 11 | 2316 | 98 | |
XM_017025173.1 | XP_016880662.1 | 11 - 12 | 2418 | 98 | |
XM_017025174.1 | XP_016880663.1 | 9 - 10 | 1858 | 98 | |
GenBank mRNA | AB051471.1 | - | 4 - 5 | 763 | 98 |
AB127405.1 | - | 10 - 11 | 2251 | 98 | |
AK126665.1 | - | 6 - 7 | 1961 | 98 | |
AK126676.1 | - | 6 - 7 | 1388 | 98 | |
AK295027.1 | - | 9 - 10 | 1858 | 98 |