The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018140.3 | NP_060610.2 | 1 - 2 | 160 | 86 |
XM_011514063.1 | XP_011512365.1 | 1 - 2 | 106 | 86 | |
XM_017009626.1 | XP_016865115.1 | 1 - 2 | 103 | 86 | |
GenBank mRNA | AB040952.1 | - | 1 - 2 | 98 | 86 |
AK001427.1 | - | 1 - 2 | 104 | 86 | |
AK299072.1 | - | 1 - 2 | 93 | 86 | |
BC000132.2 | - | 1 - 2 | 99 | 86 | |
BC001750.1 | - | 1 - 2 | 86 | 86 | |
BG718176.1 | - | 1 - 2 | 182 | 86 |