The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190274.1 | NP_001177203.1 | 15 - 16 | 1991 | 73 |
NM_025133.4 | NP_079409.3 | 15 - 16 | 1780 | 73 | |
XM_005264572.4 | XP_005264629.1 | 15 - 16 | 1985 | 73 | |
XM_005264573.4 | XP_005264630.1 | 15 - 16 | 1938 | 73 | |
XM_017005015.1 | XP_016860504.1 | 15 - 16 | 1938 | 73 | |
XM_017005016.1 | XP_016860505.1 | 15 - 16 | 1804 | 73 | |
XM_017005017.1 | XP_016860506.1 | 15 - 16 | 1804 | 73 | |
GenBank mRNA | AF351618.1 | - | 15 - 16 | 1793 | 73 |
AK022735.1 | - | 11 - 12 | 1293 | 73 | |
AK097734.1 | - | 4 - 5 | 528 | 73 | |
AK292877.1 | - | 15 - 16 | 1830 | 73 | |
AK304140.1 | - | 15 - 16 | 1695 | 73 | |
AK315108.1 | - | 15 - 16 | 1780 | 73 | |
AY827075.1 | - | 15 - 16 | 1918 | 73 | |
BC012728.2 | - | 1 - 2 | 64 | 73 | |
BC043258.1 | - | 15 - 16 | 1882 | 73 | |
BC130445.1 | - | 15 - 16 | 1716 | 73 | |
BC136480.1 | - | 15 - 16 | 1716 | 73 |